tert-Butyl 3-(pyrazin-2-yl)azetidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A1249292-100MG A1249292-1G A1249292-250MG
76680-230EA 72.15 USD
76680-230 76680-232 76680-234
tert-Butyl 3-(pyrazin-2-yl)azetidine-1-carboxylate 97%
tert-Butyl 3-(pyrazin-2-yl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₇N₃O₂
MW: 235.2823 g/mol
MDL Number: MFCD20528746
CAS Number: 1146085-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR