2,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole 98%

Supplier: Ambeed
Warning

A1249008-100MG A1249008-1G A1249008-250MG
76680-216EA 74 USD
76680-216 76680-218 76680-220
2,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole 98%
2,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

Precaución: for Research Use Only

Formula: C₁₅H₂₁BN₂O₂
MW: 272.1504 g/mol
MDL Number: MFCD12028666
CAS Number: 1315335-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR