N1,N2-Bis(2,4,6-trimethoxyphenyl)ethanediamide 97% (by HPLC)

Supplier: Ambeed
Danger

A1246501-100MG A1246501-1G A1246501-250MG A1246501-5G
76680-194EA 25 USD
76680-194 76680-196 76680-198 76680-200
N1,N2-Bis(2,4,6-trimethoxyphenyl)ethanediamide 97% (by HPLC)
N1,N2-Bis(2,4,6-trimethoxyphenyl)ethanediamide

Precaución: for Research Use Only

Formula: C₂₀H₂₄N₂O₈
MDL Number: MFCD30491289
CAS Number: 957476-07-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR