Ethyl 2-methylene-5-oxohexahydro-1H-pyrrolizine-7a-carboxylate 98%

Supplier: Ambeed
Warning

A1246412-100MG A1246412-250MG A1246412-1G A1246412-5G
76680-190EA 66.84 USD
76680-190 76680-192 77549-474 77571-484
Ethyl 2-methylene-5-oxohexahydro-1H-pyrrolizine-7a-carboxylate 98%
Ethyl 2-methylene-5-oxohexahydro-1H-pyrrolizine-7a-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.2417 g/mol
CAS Number: 942603-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR