3-(6-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione 98%

Supplier: Ambeed
Warning

A1246330-250MG A1246330-1G A1246330-5G A1246330-25G
76680-184EA 13 USD
76680-184 77549-470 77549-472 77566-394
3-(6-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione 98%
3-(6-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Precaución: For research use only.

Formula: C₁₃H₁₁BrN₂O₃
MDL Number: MFCD32667217
CAS Number: 2304513-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR