4-Bromo-6-chloro-2-methylpyridin-3-amine 98%

Supplier: Ambeed
Warning

A1246193-100MG A1246193-1G A1246193-250MG A1246193-5G
76680-180EA 37 USD
76680-180 76680-182 77866-980 77866-981
4-Bromo-6-chloro-2-methylpyridin-3-amine 98%
4-Bromo-6-chloro-2-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₆BrClN₂
MW: 221.4822 g/mol
MDL Number: MFCD32670596
CAS Number: 1351813-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR