((2R,7aS)-2-Fluorohexahydro-1H-pyrrolizin-7a-yl)methanol 98%

Supplier: Ambeed
Warning

A1246021-100MG A1246021-250MG A1246021-5G A1246021-10G A1246021-25G A1246021-100G A1246021-1G
76680-164EA 21.22 USD
76680-164 76680-166 76680-168 77549-466 77549-468 77566-388 77566-390
((2R,7aS)-2-Fluorohexahydro-1H-pyrrolizin-7a-yl)methanol 98%
((2R,7aS)-2-Fluorohexahydro-1H-pyrrolizin-7a-yl)methanol

Precaución: For research use only.

Formula: C₈H₁₄FNO
MW: 159.2013 g/mol
MDL Number: MFCD31641130
CAS Number: 2097518-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR