Ethyl 2,5-dioxohexahydro-1H-pyrrolizine-7a-carboxylate 97%

Supplier: Ambeed
Warning

A1246015-250MG A1246015-5G A1246015-1G A1246015-100MG
76680-160EA 47.75 USD
76680-160 76680-162 77592-422 77621-212
Ethyl 2,5-dioxohexahydro-1H-pyrrolizine-7a-carboxylate 97%
Ethyl 2,5-dioxohexahydro-1H-pyrrolizine-7a-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄
MW: 211.215 g/mol
MDL Number: MFCD34165661
CAS Number: 942603-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR