2-(Hydroxymethyl)-4-iodophenol 98%
Supplier: Ambeed
|
Warning
|
A1245737-100MG
A1245737-1G
A1245737-250MG
76680-148EA
39
USD
76680-148
76680-150
76680-152
2-(Hydroxymethyl)-4-iodophenol 98%
2-(Hydroxymethyl)-4-iodophenol
Precaución: for Research Use Only
Formula:
C₇H₇IO₂ |
MDL Number:
MFCD06202813 CAS Number: 14056-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...