Zaltoprofen 99%

Supplier: Ambeed
Warning

Synonyms: 2-(10,11-Dihydro-10-oxodibenzo[b,f]thiepin-2-yl)propionic Acid, 2-(10-Oxo-10,11-dihydrodibenzo[b,f]thiepin-2-yl)propanoic acid

A1245402-1G A1245402-25G A1245402-5G
76680-142EA 12.74 USD
76680-142 76680-144 76680-146
Zaltoprofen 99%
Zaltoprofen

Precaución: for Research Use Only

Formula: C₁₇H₁₄O₃S
MW: 298.36 g/mol
MDL Number: MFCD00864323
CAS Number: 74711-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR