3-Bromo-5-chloro-tert-butylbenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-(tert-butyl)-5-chlorobenzene

A1244594-25G A1244594-1G A1244594-5G
76680-134EA 267.35 USD
76680-134 77621-208 77621-210
3-Bromo-5-chloro-tert-butylbenzene 98%
3-Bromo-5-chloro-tert-butylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrCl
MW: 247.559 g/mol
MDL Number: MFCD18089328
CAS Number: 1263377-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR