3,4-Dibromo-2-methylaniline 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dibromo-2-methyl-phenylamine

A1242150-100MG A1242150-1G A1242150-250MG
76680-120EA 34 USD
76680-120 76680-122 76680-124
3,4-Dibromo-2-methylaniline 98%
3,4-Dibromo-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇Br₂N
MDL Number: MFCD11845872
CAS Number: 858843-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR