N4-Hydroxycytidine 98%

Supplier: Ambeed
Danger

Synonyms: EIDD-1931, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-hydroxymethyl-tetrahydro-furan-2-yl)-4-hydroxyamino-1H-pyrimidin-2-one, Beta-D-N4-hydroxycytidine, NHC, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-(hydroxyimino)-3,4-dihydropyrimidin-2(1H)-one, 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-(hydroxyamino)pyrimidin-2-one, 4-oxime, Uridine

A1241627-10MG A1241627-5MG A1241627-25G
76680-116EA 9.55 USD
76680-116 76680-118 77621-206
N4-Hydroxycytidine 98%
N4-Hydroxycytidine

Precaución: for Research Use Only

Formula: C₉H₁₃N₃O₆
MW: 259.22 g/mol
MDL Number: MFCD01675695
CAS Number: 3258-02-4
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR