2-Methylsulfanyl-4-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A1241121-100MG A1241121-1G A1241121-250MG
76680-102EA 9.55 USD
76680-102 76680-104 76680-106
2-Methylsulfanyl-4-(trifluoromethyl)benzoic acid 98%
2-Methylsulfanyl-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂S
MW: 236.2109 g/mol
MDL Number: MFCD22489208
CAS Number: 142994-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR