4-Bromo-3-fluoro-2-methoxybenzonitrile 98%

Supplier: Ambeed
Warning

A1239820-100MG A1239820-1G A1239820-250MG
76680-094EA 74 USD
76680-094 76680-096 76680-098
4-Bromo-3-fluoro-2-methoxybenzonitrile 98%
4-Bromo-3-fluoro-2-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFNO
MDL Number: MFCD23707400
CAS Number: 1427452-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR