3-Chlorophenyl trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

A1239612-1G A1239612-250MG A1239612-5G
76680-088EA 71.09 USD
76680-088 76680-090 76680-092
3-Chlorophenyl trifluoromethanesulfonate 98%
3-Chlorophenyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O₃S
MW: 260.6181 g/mol
MDL Number: MFCD25541794
CAS Number: 86364-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR