4-(Trifluoromethyl)phenyl trifluoromethanesulfonate 98%

Supplier: Ambeed
Danger

A1239168-1G A1239168-250MG A1239168-25G A1239168-5G
76680-072EA 95.49 USD
76680-072 76680-074 76680-076 76680-078
4-(Trifluoromethyl)phenyl trifluoromethanesulfonate 98%
4-(Trifluoromethyl)phenyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₈H₄F₆O₃S
MW: 294.1710 g/mol
MDL Number: MFCD07784330
CAS Number: 146397-87-7
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR