3-Chloro-2-fluoro-6-(trifluoromethyl)benzyl bromide 98%

Supplier: Ambeed
Danger

A1237255-100MG A1237255-1G A1237255-250MG
76680-066EA 254.62 USD
76680-066 76680-068 76680-070
3-Chloro-2-fluoro-6-(trifluoromethyl)benzyl bromide 98%
3-Chloro-2-fluoro-6-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₈H₄BrClF₄
MW: 291.4680 g/mol
MDL Number: MFCD01631346
CAS Number: 261763-10-4
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR