3-Bromo-4-fluorothioanisole 98%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-4-fluorophenyl)(methyl)sulphane

A1237250-100MG A1237250-1G
76680-062EA 63 USD
76680-062 76680-064
3-Bromo-4-fluorothioanisole 98%
3-Bromo-4-fluorothioanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFS
MDL Number: MFCD22488902
CAS Number: 1511032-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR