4-Bromo-1-(phenylsulfonyl)-1H-indole 98%

Supplier: Ambeed
Warning

A1237053-1G A1237053-250MG
76680-058EA 64.72 USD
76680-058 76680-060
4-Bromo-1-(phenylsulfonyl)-1H-indole 98%
4-Bromo-1-(phenylsulfonyl)-1H-indole

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrNO₂S
MW: 336.2037 g/mol
MDL Number: MFCD12828204
CAS Number: 412048-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR