2,6-Dihydroxyterephthalic acid 98%

Supplier: Ambeed
Warning

A1235626-1G A1235626-5G A1235626-25G
76680-052EA 20.16 USD
76680-052 76680-054 77566-360
2,6-Dihydroxyterephthalic acid 98%
2,6-Dihydroxyterephthalic acid

Precaución: For research use only.

Formula: C₈H₆O₆
MW: 198.1296 g/mol
MDL Number: MFCD01570246
CAS Number: 69660-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR