1-Ethyl-4-isobutylbenzene 97%
Supplier: Ambeed
|
Warning
|
A1235118-100MG
A1235118-1G
A1235118-250MG
76680-042EA
47.75
USD
76680-042
76680-044
76680-046
1-Ethyl-4-isobutylbenzene 97%
1-Ethyl-4-isobutylbenzene
Precaución: for Research Use Only
Formula:
C₁₂H₁₈ MW: 162.2713 g/mol |
MDL Number:
MFCD08275587 CAS Number: 100319-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...