(2S,4R)-1-((S)-3,3-Dimethyl-2-(2-(2-(4-(6-((4-(trifluoromethoxy)phenyl)amino)pyrimidin-4-yl)phenoxy)ethoxy)acetamido)butanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: GMB-475

A1235114-10MG A1235114-1MG A1235114-25MG A1235114-50MG A1235114-5MG
76680-030EA 127.31 USD
76680-030 76680-032 76680-034 76680-036 76680-038
(2S,4R)-1-((S)-3,3-Dimethyl-2-(2-(2-(4-(6-((4-(trifluoromethoxy)phenyl)amino)pyrimidin-4-yl)phenoxy)ethoxy)acetamido)butanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide 98%
(2S,4R)-1-((S)-3,3-Dimethyl-2-(2-(2-(4-(6-((4-(trifluoromethoxy)phenyl)amino)pyrimidin-4-yl)phenoxy)ethoxy)acetamido)butanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₄₃H₄₆F₃N₇O₇S
MW: 861.9283 g/mol
MDL Number: MFCD32220458
CAS Number: 2490599-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR