N-(4-(4-((5-(2-Methoxyethoxy)pyrazin-2-yl)thio)-2,6-dimethylphenyl)thiazol-2-yl)isonicotinamide 4-methylbenzenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: T-1101 tosylate

A1235104-10MG A1235104-1MG A1235104-25MG A1235104-50MG A1235104-5MG A1235104-100MG A1235104-250MG
76680-020EA 83.82 USD
76680-020 76680-022 76680-024 76680-026 76680-028 77621-196 77621-198
N-(4-(4-((5-(2-Methoxyethoxy)pyrazin-2-yl)thio)-2,6-dimethylphenyl)thiazol-2-yl)isonicotinamide 4-methylbenzenesulfonate 98%
N-(4-(4-((5-(2-Methoxyethoxy)pyrazin-2-yl)thio)-2,6-dimethylphenyl)thiazol-2-yl)isonicotinamide 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₃₁H₃₁N₅O₆S₃
MW: 665.8 g/mol
MDL Number: MFCD32689635
CAS Number: 2250404-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR