tert-Butyl 4-fluoro-1-oxo-1,3-dihydrospiro[indene-2,4'-piperidine]-1'-carboxylate 98%

Supplier: Ambeed
Warning

A1235000-100MG A1235000-1G A1235000-250MG
76680-008EA 128.37 USD
76680-008 76680-010 76680-012
tert-Butyl 4-fluoro-1-oxo-1,3-dihydrospiro[indene-2,4'-piperidine]-1'-carboxylate 98%
tert-Butyl 4-fluoro-1-oxo-1,3-dihydrospiro[indene-2,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₂FNO₃
MW: 319.3706 g/mol
MDL Number: MFCD32876781
CAS Number: 2377354-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR