tert-Butyl 6-bromo-1-oxo-1,3-dihydrospiro[indene-2,4'-piperidine]-1'-carboxylate 98%

Supplier: Ambeed
Warning

A1234992-100MG A1234992-250MG A1234992-1G
76680-004EA 66.84 USD
76680-004 76680-006 77621-194
tert-Butyl 6-bromo-1-oxo-1,3-dihydrospiro[indene-2,4'-piperidine]-1'-carboxylate 98%
tert-Butyl 6-bromo-1-oxo-1,3-dihydrospiro[indene-2,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₂BrNO₃
MW: 380.2762 g/mol
MDL Number: MFCD33401648
CAS Number: 2245084-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR