Methyl 2-cyano-3-(5-(4-sulfamoylphenyl)furan-2-yl)acrylate 98%

Supplier: Ambeed
Warning

A1234954-10MG A1234954-1MG A1234954-25MG A1234954-50MG A1234954-5MG
76679-994EA 40 USD
76679-994 76679-996 76679-998 76680-000 76680-002
Methyl 2-cyano-3-(5-(4-sulfamoylphenyl)furan-2-yl)acrylate 98%
Methyl 2-cyano-3-(5-(4-sulfamoylphenyl)furan-2-yl)acrylate

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂O₅S
MDL Number: MFCD01847328
CAS Number: 292053-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR