Sodium 2-(acetylthio)ethanesulfonate 97%

Supplier: Ambeed
Warning

A1234768-1G A1234768-250MG
76679-990EA 105 USD
76679-990 76679-992
Sodium 2-(acetylthio)ethanesulfonate 97%
Sodium 2-(acetylthio)ethanesulfonate

Precaución: for Research Use Only

Formula: C₄H₇NaO₄S₂
MDL Number: MFCD18379157
CAS Number: 76274-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR