(5-Chloro-2-methyl-3-nitrophenyl)methanol 98%

Supplier: Ambeed
Warning

A1234370-100MG A1234370-1G A1234370-250MG A1234370-5G
76679-980EA 23 USD
76679-980 76679-982 76679-984 76679-986
(5-Chloro-2-methyl-3-nitrophenyl)methanol 98%
(5-Chloro-2-methyl-3-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MDL Number: MFCD28756978
CAS Number: 1613329-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR