7-Bromo-2,4-dichloro-8-fluoro-quinazoline 97%

Supplier: Ambeed
Warning

A1233225-10G A1233225-1G A1233225-5G A1233225-25G A1233225-100MG A1233225-250MG
76679-972EA 197 USD
76679-972 76679-974 76679-976 77571-428 77866-925 77866-926
7-Bromo-2,4-dichloro-8-fluoro-quinazoline 97%
7-Bromo-2,4-dichloro-8-fluoro-quinazoline

Precaución: for Research Use Only

Formula: C₈H₂BrCl₂FN₂
MW: 295.9233 g/mol
MDL Number: MFCD32203041
CAS Number: 2168499-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR