Chlorobis(cyclooctene)iridium(I) dimer 98+%

Supplier: Ambeed
Warning

A1233163-100MG A1233163-1G A1233163-250MG A1233163-5G
76679-962EA 71 USD
76679-962 76679-964 76679-966 76679-968 76679-970
Chlorobis(cyclooctene)iridium(I) dimer 98+%
Chlorobis(cyclooctene)iridium(I) dimer

Precaución: for Research Use Only

Formula: C₃₂H₅₆Cl₂Ir₂
MDL Number: MFCD00213465
CAS Number: 12246-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR