Methoxy(cyclooctadiene)iridium(I) dimer 98% + (Carbon 31.3-33.9%)

Supplier: Ambeed
Warning

Synonyms: Di-mu-methoxobis(1,5-cyclooctadiene)diiridium(I), Bis(1,5-cyclooctadiene)di-mu-methoxydiiridium(I), (1,5-Cyclooctadiene)(methoxy)iridium(I) dimer, Bis(1,5-cyclooctadiene)dimethoxydiiridium

A1233159-250MG A1233159-5G
76679-954EA 286.45 USD
76679-954 76679-956 76679-958
Methoxy(cyclooctadiene)iridium(I) dimer 98% + (Carbon 31.3-33.9%)
Methoxy(cyclooctadiene)iridium(I) dimer

Precaución: for Research Use Only

Formula: C₁₈H₃₀Ir₂O₂
MW: 662.87 g/mol
MDL Number: MFCD08459360
CAS Number: 12148-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR