(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer 98% (Carbon 28.8-31.5%)

Supplier: Ambeed
Danger

A1233157-100MG A1233157-1G A1233157-250MG A1233157-5G
76679-946EA 61 USD
76679-946 76679-948 76679-950 76679-952
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer 98% (Carbon 28.8-31.5%)
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer

Precaución: for Research Use Only

Formula: C₂₀H₃₀Cl₄Ir₂
MDL Number: MFCD00075435
CAS Number: 12354-84-6
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR