Bis(dibenzylideneacetone)palladium 98+%

Supplier: Ambeed
Warning

Synonyms: Bis(dibenzylideneacetone)palladium(0)

A1233151-100MG A1233151-250MG
76679-934EA 23.34 USD
76679-934 76679-936 76679-938
Bis(dibenzylideneacetone)palladium 98+%
Bis(dibenzylideneacetone)palladium

Precaución: for Research Use Only

Formula: C₃₄H₂₈O₂Pd
MW: 575 g/mol
MDL Number: MFCD00051942
CAS Number: 32005-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR