Palladium(II) acetylacetonate 99%
Supplier: Ambeed
|
Warning
|
A1233143-100MG
A1233143-1G
A1233143-250MG
A1233143-5G
76679-926EA
29
USD
76679-926
76679-928
76679-930
76679-932
Palladium(II) acetylacetonate 99%
Palladium(II) acetylacetonate
Precaución: for Research Use Only
Formula:
C₁₀H₁₄O₄Pd |
MDL Number:
MFCD00000025 CAS Number: 14024-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...