Palladium(II) trifluoroacetate 98+%

Supplier: Ambeed
Warning

Synonyms: Palladium(II) 2,2,2-trifluoroacetate

A1233133-100MG A1233133-1G A1233133-250MG
76679-920EA 27.59 USD
76679-920 76679-922 76679-924
Palladium(II) trifluoroacetate 98+%
Palladium(II) trifluoroacetate

Precaución: for Research Use Only

Formula: C₄F₆O₄Pd
MW: 332.45 g/mol
MDL Number: MFCD00013204
CAS Number: 42196-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR