Chloro(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)[2-(2-aminoethylphenyl)]palladium(II) 98+%
Supplier: Ambeed
|
Warning
|
A1233123-100MG
A1233123-250MG
A1233123-5G
76679-910EA
18.04
USD
76679-910
76679-912
77621-178
Chloro(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)[2-(2-aminoethylphenyl)]palladium(II) 98+%
Chloro(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)[2-(2-aminoethylphenyl)]palladium(II)
Precaución: for Research Use Only
Formula:
C₃₄H₄₅ClNO₂PPd MW: 672.57 g/mol |
MDL Number:
MFCD15144765 CAS Number: 1028206-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...