Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Dicyclohexylphosphino-2',4',6'-triisopropyl-1,1'-biphenyl)[2-(2'-amino-1,1'-biphenyl)]palladium(ii) methanesulphonate
A1233103-1G
A1233103-250MG
A1233103-5G
A1233103-10G
A1233103-25G
76679-894EA
26
USD
76679-894
76679-896
76679-898
77592-404
77596-506
Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98+%
Methanesulfonato(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II)
Precaución: for Research Use Only
Formula:
C₄₆H₆₂NO₃PPdS |
MDL Number:
MFCD22417234 CAS Number: 1445085-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...