Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98+%

Supplier: Ambeed
Warning

Synonyms: RuPhos-Pd-G3

A1233095-100MG A1233095-250MG
76679-890EA 11.67 USD
76679-890 76679-892
Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98+%
Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₄₃H₅₆NO₅PPdS
MW: 836.37 g/mol
MDL Number: MFCD22572684
CAS Number: 1445085-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR