(2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl)[2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate 98%

Supplier: Ambeed
Warning

A1233091-100MG A1233091-1G A1233091-250MG
76679-884EA 13 USD
76679-884 76679-886 76679-888
(2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl)[2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate 98%
(2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl)[2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate

Precaución: for Research Use Only

Formula: C₄₀H₅₁NO₈PPdS₂
MW: 875.36 g/mol
MDL Number: MFCD29472404
CAS Number: 1445085-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR