Rhodium(II) trifluoroacetate dimer 98%+ (Carbon: 14.61 ± 1.40%)

Supplier: Ambeed
Warning

Synonyms: Trifluoroacetic acid rhodium(II) salt dimer

A1233089-100MG A1233089-250MG A1233089-1G
76679-880EA 95 USD
76679-880 76679-882 77621-176
Rhodium(II) trifluoroacetate dimer 98%+ (Carbon: 14.61 ± 1.40%)
Rhodium(II) trifluoroacetate dimer

Precaución: for Research Use Only

Formula: C₈F₁₂O₈Rh₂
MDL Number: MFCD00209611
CAS Number: 31126-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR