Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimer 98+%

Supplier: Ambeed
Warning

A1233085-100MG A1233085-250MG
76679-876EA 164 USD
76679-876 76679-878
Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimer 98+%
Bicyclo[2.2.1]hepta-2,5-diene-rhodium(I) chloride dimer

Precaución: for Research Use Only

Formula: C₁₄H₈Cl₂Rh₂
MDL Number: MFCD00198060
CAS Number: 12257-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR