Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer 97%

Supplier: Ambeed
Warning

A1233059-100MG A1233059-250MG A1233059-50MG
76679-866EA 169 USD
76679-866 76679-868 76679-870
Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer 97%
Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer

Precaución: for Research Use Only

Formula: C₁₆H₂₆O₂Rh₂
MDL Number: MFCD04974244
CAS Number: 73468-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR