[(2-Di-tert-butylphosphino-3,6-dimethoxy-2',4',6'-triisopropyl-1,1'-biphenyl)-2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: tBuBrettPhos Pd G3

A1233055-100MG A1233055-250MG A1233055-5G A1233055-1G A1233055-25G
76679-862EA 16.98 USD
76679-862 76679-864 77549-450 77549-448 77571-426
[(2-Di-tert-butylphosphino-3,6-dimethoxy-2',4',6'-triisopropyl-1,1'-biphenyl)-2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate 98%
[(2-Di-tert-butylphosphino-3,6-dimethoxy-2',4',6'-triisopropyl-1,1'-biphenyl)-2-(2'-amino-1,1'-biphenyl)]palladium(II) methanesulfonate

Precaución: for Research Use Only

Formula: C₄₄H₆₂NO₅PPdS
MW: 854.43 g/mol
MDL Number: MFCD25976528
CAS Number: 1536473-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR