Bis(cyclopentadienyl)ruthenium 99+%

Supplier: Ambeed
Warning

Synonyms: Ruthenocene, Bis(cyclopentadienyl)ruthenium(II)

A1233053-100MG A1233053-1G A1233053-250MG A1233053-5G
76679-854EA 20 USD
76679-854 76679-856 76679-858 76679-860
Bis(cyclopentadienyl)ruthenium 99+%
Bis(cyclopentadienyl)ruthenium

Precaución: for Research Use Only

Formula: C₁₀Ru
MW: 231.26
MDL Number: MFCD00001442
CAS Number: 1287-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR