Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate 98+%

Supplier: Ambeed
Warning

Synonyms: Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate

A1233037-1G A1233037-250MG
76679-850EA 235 USD
76679-850 76679-852
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate 98+%
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₇H₂₄F₃O₃RhS
MW: 468.34 g/mol
MDL Number: MFCD00143752
CAS Number: 99326-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR