Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate 98+%

Supplier: Ambeed
Danger

A1233031-100MG A1233031-250MG
76679-840EA 55 USD
76679-840 76679-842
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate 98+%
Bis(1,5-cyclooctadiene)rhodium (I) tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₆H₂₄BF₄Rh
MDL Number: MFCD00075045
CAS Number: 35138-22-8
UN: 2921
ADR: 8,4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR