Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98+%

Supplier: Ambeed
Warning

Synonyms: (1,5-Cyclooctadiene)rhodium chloride dimer

A1233027-100MG A1233027-1G A1233027-250MG
76679-834EA 72.15 USD
76679-834 76679-836 76679-838
Chloro(1,5-cyclooctadiene)rhodium(I) dimer 98+%
Chloro(1,5-cyclooctadiene)rhodium (I) dimer

Precaución: for Research Use Only

Formula: C₁₆H₂₄Cl₂Rh₂
MW: 493.08 g/mol
MDL Number: MFCD00012415
CAS Number: 12092-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR