(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer 98% (Carbon: 38.9 ± 0.8%)

Supplier: Ambeed
Warning

Synonyms: Dichloro(pentamethylcyclopentadienyl)rhodium (III) dimer, [Rh(Cp-)Cl2]2, Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, Dichloro(pentamethylcyclopentadienyl)rhodium(III) Dimer

A1233023-100MG A1233023-1G A1233023-250MG A1233023-50MG
76679-824EA 99.73 USD
76679-824 76679-826 76679-828 76679-830
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer 98% (Carbon: 38.9 ± 0.8%)
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer

Precaución: for Research Use Only

Formula: C₂₀H₃₀Cl₄Rh₂
MW: 618.07 g/mol
MDL Number: MFCD00061552
CAS Number: 12354-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR