(Triphenylphosphine)gold(I) chloride 98+%

Supplier: Ambeed
Warning

Synonyms: Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold

A1233009-100MG A1233009-10G A1233009-250MG A1233009-5G
76679-810EA 20 USD
76679-810 76679-812 76679-814 76679-816 76679-818
(Triphenylphosphine)gold(I) chloride 98+%
(Triphenylphosphine)gold(I) chloride

Precaución: for Research Use Only

Formula: C₁₈H₁₅AuClP
MW: 494.71
MDL Number: MFCD00009588
CAS Number: 14243-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR